(3-Cyclopropyl-4-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

48791-500MG 48791-1G 48791-5G 48791-25G
77327-698EA 206 USD
77327-698 77351-276 77364-028 77386-446
(3-Cyclopropyl-4-fluorophenyl)methanol ≥97%
(3-Cyclopropyl-4-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO
MDL Number: MFCD28402849
CAS Number: 1429421-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR