(3-(Benzyloxy)-2,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48782-500MG 48782-1G 48782-5G 48782-25G
77327-694EA 176.13 USD
77327-694 77346-526 77364-024 77386-442
(3-(Benzyloxy)-2,4-difluorophenyl)methanol ≥97%
(3-(Benzyloxy)-2,4-difluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD33403907
CAS Number: 2635937-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR