Methyl 3-(benzyloxy)-2,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48780-500MG 48780-1G 48780-5G 48780-25G
77327-690EA 170.98 USD
77327-690 77346-372 77364-020 77386-438
Methyl 3-(benzyloxy)-2,4-difluorobenzoate ≥97%
Methyl 3-(benzyloxy)-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD33403905
CAS Number: 2635937-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR