3-(Benzyloxy)-2,4-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48779-500MG 48779-1G 48779-5G 48779-25G
77327-688EA 100.94 USD
77327-688 77342-918 77364-018 77375-556
3-(Benzyloxy)-2,4-difluorobenzoic acid ≥97%
3-(Benzyloxy)-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD32902763
CAS Number: 1637224-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR