2-Isopropoxy-4-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48773-500MG 48773-1G 48773-5G 48773-25G
77327-686EA 122.57 USD
77327-686 77343-914 77364-016 77375-554
2-Isopropoxy-4-(methoxymethoxy)benzaldehyde ≥97%
2-Isopropoxy-4-(methoxymethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902761
CAS Number: 1165713-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR