(4-Ethoxy-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48771-500MG 48771-1G 48771-5G 48771-25G
77327-684EA 221.45 USD
77327-684 77351-274 77364-014 77386-436
(4-Ethoxy-2-methoxyphenyl)methanol ≥97%
(4-Ethoxy-2-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26399197
CAS Number: 181819-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR