4-Ethoxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48770-500MG 48770-1G 48770-5G 48770-25G
77327-682EA 138.02 USD
77327-682 77344-648 77364-012 77386-434
4-Ethoxy-2-methoxybenzaldehyde ≥97%
4-Ethoxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24388335
CAS Number: 42924-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR