1,2-bis(2-fluoropyridin-4-yl)ethane ≥97%

Supplier: AOBChem USA

Warning

48720-500MG 48720-1G 48720-5G 48720-25G
77327-676EA 210.12 USD
77327-676 77351-268 77364-004 77386-428
1,2-bis(2-fluoropyridin-4-yl)ethane ≥97%
1,2-bis(2-fluoropyridin-4-yl)ethane

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD28338361
CAS Number: 954097-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR