2,5-Dibromo-1-ethyl-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48714-500MG 48714-1G 48714-5G 48714-25G
77327-674EA 214.24 USD
77327-674 77351-266 77364-002 77386-426
2,5-Dibromo-1-ethyl-3-methylbenzene ≥97%
2,5-Dibromo-1-ethyl-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂
MW: 277.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547779
CAS Number: 1208075-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR