5-Bromo-2-chloro-1-ethyl-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48713-500MG 48713-1G 48713-5G 48713-25G
77327-672EA 229.69 USD
77327-672 77351-264 77364-000 77386-424
5-Bromo-2-chloro-1-ethyl-3-methylbenzene ≥97%
5-Bromo-2-chloro-1-ethyl-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrCl
MW: 233.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529985
CAS Number: 2710785-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR