2-(Benzyloxy)-3-chloro-5-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

48707-500MG 48707-1G 48707-5G 48707-25G
77327-670EA 99.91 USD
77327-670 77342-802 77363-998 77375-548
2-(Benzyloxy)-3-chloro-5-fluorobenzoic acid ≥95%
2-(Benzyloxy)-3-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₃
MW: 280.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756468
CAS Number: 2807447-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR