2-Chloro-6-iodo-3-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
48699-500MG
48699-1G
48699-5G
48699-25G
77327-664EA
202.91
USD
77327-664
77351-260
77363-992
77386-418
2-Chloro-6-iodo-3-methoxybenzoic acid ≥97%
2-Chloro-6-iodo-3-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆ClIO₃ MW: 312.489 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34756464 CAS Number: 2385768-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...