2-Chloro-6-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48697-500MG 48697-1G 48697-5G 48697-25G
77327-662EA 168.92 USD
77327-662 77346-258 77363-990 77386-416
2-Chloro-6-iodo-3-methoxybenzaldehyde ≥97%
2-Chloro-6-iodo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClIO₂
MW: 296.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403901
CAS Number: 2383017-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR