Methyl 3,6-dichloro-2-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48695-500MG 48695-1G 48695-5G 48695-25G
77327-660EA 150.38 USD
77327-660 77345-342 77363-988 77386-414
Methyl 3,6-dichloro-2-hydroxybenzoate ≥97%
Methyl 3,6-dichloro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28786469
CAS Number: 53066-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR