1,4-Dichloro-2-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

48687-500MG 48687-1G 48687-5G 48687-25G
77327-658EA 281.19 USD
77327-658 77351-258 77363-986 77386-412
1,4-Dichloro-2-methoxy-3-methylbenzene ≥95%
1,4-Dichloro-2-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MW: 191.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786751
CAS Number: 1803835-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR