3-Bromo-6-chloro-2-fluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48670-500MG 48670-1G 48670-5G 48670-25G
77327-656EA 214.24 USD
77327-656 77351-256 77363-984 77386-410
3-Bromo-6-chloro-2-fluoro-N,N-dimethylbenzamide ≥97%
3-Bromo-6-chloro-2-fluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD32902757
CAS Number: 2586126-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR