3-Bromo-6-chloro-2-fluoro-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

48669-500MG 48669-1G 48669-5G 48669-25G
77327-654EA 272.95 USD
77327-654 77351-254 77363-982 77386-408
3-Bromo-6-chloro-2-fluoro-N-methylbenzamide ≥95%
3-Bromo-6-chloro-2-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD32902756
CAS Number: 2504204-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR