3-Bromo-6-chloro-N-cyclopropyl-2-fluorobenzamide ≥95%

Supplier: AOBChem USA

Warning

48668-500MG 48668-1G 48668-25G 48668-5G
77327-652EA 299.73 USD
77327-652 77351-252 77386-406 77375-546
3-Bromo-6-chloro-N-cyclopropyl-2-fluorobenzamide ≥95%
3-Bromo-6-chloro-N-cyclopropyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈BrClFNO
MW: 292.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902755
CAS Number: 2504201-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR