(3-Bromo-6-chloro-2-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48666-500MG 48666-1G 48666-5G 48666-25G
77327-650EA 214.24 USD
77327-650 77351-250 77363-980 77386-404
(3-Bromo-6-chloro-2-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%
(3-Bromo-6-chloro-2-fluorophenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD32902753
CAS Number: 2586126-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR