(3-Bromo-6-chloro-2-fluorophenyl)(piperidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48665-500MG 48665-1G 48665-25G 48665-5G
77327-648EA 299.73 USD
77327-648 77351-248 77386-402 77375-544
(3-Bromo-6-chloro-2-fluorophenyl)(piperidin-1-yl)methanone ≥95%
(3-Bromo-6-chloro-2-fluorophenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrClFNO
MW: 320.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902752
CAS Number: 2504203-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR