(3-Bromo-6-chloro-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48662-500MG 48662-1G 48662-25G 48662-5G
77327-646EA 299.73 USD
77327-646 77351-246 77386-400 77375-542
(3-Bromo-6-chloro-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3-Bromo-6-chloro-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrClFN₂O
MW: 335.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902749
CAS Number: 2504202-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR