1,3-Dichloro-2-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

48657-500MG 48657-1G 48657-5G
77338-000EA 457.32 USD
77338-000 77351-244 77375-540
1,3-Dichloro-2-ethylbenzene ≥95%
1,3-Dichloro-2-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18968150
CAS Number: 33407-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR