2,4,6-trifluoro-3-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48651-500MG 48651-1G 48651-5G
77338-218EA 469.68 USD
77338-218 77351-242 77375-538
2,4,6-trifluoro-3-hydroxybenzoic acid ≥97%
2,4,6-trifluoro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28800136
CAS Number: 91659-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR