4-(Benzyloxy)-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48641-500MG 48641-1G 48641-5G 48641-25G
77327-644EA 136.99 USD
77327-644 77344-530 77363-978 77386-398
4-(Benzyloxy)-3-(trifluoromethyl)benzaldehyde ≥97%
4-(Benzyloxy)-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₂
MW: 280.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18330920
CAS Number: 477980-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR