Ethyl 3-chloro-4-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48638-500MG 48638-1G 48638-5G 48638-25G
77327-642EA 97.85 USD
77327-642 77342-666 77363-976 77375-536
Ethyl 3-chloro-4-propoxybenzoate ≥97%
Ethyl 3-chloro-4-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34756457
CAS Number: 860697-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR