1-Chloro-2,3-difluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48599-500MG 48599-1G 48599-5G 48599-25G
77327-634EA 178.19 USD
77327-634 77346-606 77363-968 77386-392
1-Chloro-2,3-difluoro-4-methylbenzene ≥97%
1-Chloro-2,3-difluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClF₂
MW: 162.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415654
CAS Number: 1261737-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR