2,3-Difluoro-N,4-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48597-1G 48597-5G 48597-25G
77341-326EA 82.4 USD
77341-326 77363-966 77375-528
2,3-Difluoro-N,4-dimethylbenzamide ≥97%
2,3-Difluoro-N,4-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34564124
CAS Number: 2734772-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR