N-Cyclopropyl-2,3-difluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48596-1G 48596-5G 48596-25G
77341-760EA 104.03 USD
77341-760 77363-964 77375-526
N-Cyclopropyl-2,3-difluoro-4-methylbenzamide ≥97%
N-Cyclopropyl-2,3-difluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD05991086
CAS Number: 708223-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR