N-Cyclohexyl-2,3-difluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48595-1G 48595-5G 48595-25G
77341-482EA 89.61 USD
77341-482 77363-962 77375-524
N-Cyclohexyl-2,3-difluoro-4-methylbenzamide ≥97%
N-Cyclohexyl-2,3-difluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34564123
CAS Number: 2734774-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR