2,3-Difluoro-N-isopropyl-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48594-1G 48594-5G 48594-25G
77341-324EA 82.4 USD
77341-324 77363-960 77375-522
2,3-Difluoro-N-isopropyl-4-methylbenzamide ≥97%
2,3-Difluoro-N-isopropyl-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34564122
CAS Number: 2734774-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR