2,3-Difluoro-N,N,4-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48593-1G 48593-5G 48593-25G
77341-322EA 82.4 USD
77341-322 77363-958 77375-520
2,3-Difluoro-N,N,4-trimethylbenzamide ≥97%
2,3-Difluoro-N,N,4-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34564121
CAS Number: 2734774-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR