(2,3-difluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48592-500MG 48592-1G 48592-5G 48592-25G
77327-632EA 126.69 USD
77327-632 77344-114 77363-956 77375-518
(2,3-difluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
(2,3-difluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO
MW: 225.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05989156
CAS Number: 701289-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR