(2,3-difluoro-4-methylphenyl)(piperidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
48591-500MG
48591-1G
48591-5G
48591-25G
77327-630EA
133.9
USD
77327-630
77344-412
77363-954
77386-390
(2,3-difluoro-4-methylphenyl)(piperidin-1-yl)methanone ≥97%
(2,3-difluoro-4-methylphenyl)(piperidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₃H₁₅F₂NO MW: 239.261 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34564120 CAS Number: 2734774-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...