(2,3-difluoro-4-methylphenyl)(morpholino)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
48590-500MG
48590-1G
48590-5G
48590-25G
77327-628EA
124.63
USD
77327-628
77344-052
77363-952
77375-516
(2,3-difluoro-4-methylphenyl)(morpholino)methanone ≥97%
(2,3-difluoro-4-methylphenyl)(morpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₃F₂NO₂ MW: 241.234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05991273 CAS Number: 708229-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...