(2,3-difluoro-4-methylphenyl)(thiomorpholino)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
48589-500MG
48589-1G
48589-5G
48589-25G
77327-626EA
124.63
USD
77327-626
77344-050
77363-950
77375-514
(2,3-difluoro-4-methylphenyl)(thiomorpholino)methanone ≥97%
(2,3-difluoro-4-methylphenyl)(thiomorpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₃F₂NOS MW: 257.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34564119 CAS Number: 2734774-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...