(2,3-difluoro-4-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48588-500MG 48588-1G 48588-5G 48588-25G
77327-624EA 124.63 USD
77327-624 77344-048 77363-948 77375-512
(2,3-difluoro-4-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2,3-difluoro-4-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₂N₂O
MW: 254.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05989589
CAS Number: 701940-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR