2-Ethoxy-5-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

48583-500MG 48583-1G 48583-5G 48583-25G
77327-622EA 121.54 USD
77327-622 77343-866 77363-946 77375-510
2-Ethoxy-5-methylbenzonitrile ≥97%
2-Ethoxy-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD14534394
CAS Number: 548784-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR