(2-Bromo-3-ethoxy-6-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48581-500MG 48581-1G 48581-5G 48581-25G
77327-620EA 136.99 USD
77327-620 77344-590 77363-944 77386-388
(2-Bromo-3-ethoxy-6-fluorophenyl)methanol ≥97%
(2-Bromo-3-ethoxy-6-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34178544
CAS Number: 2385171-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR