Methyl 3-fluoro-2-methoxy-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

48568-500MG 48568-1G 48568-5G
77327-612EA 334.75 USD
77327-612 77351-224 77375-502
Methyl 3-fluoro-2-methoxy-4-methylbenzoate ≥95%
Methyl 3-fluoro-2-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756449
CAS Number: 2116892-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR