Ethyl 3-fluoro-2-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48567-500MG 48567-1G 48567-5G
77327-610EA 345.05 USD
77327-610 77351-222 77375-500
Ethyl 3-fluoro-2-methoxy-4-methylbenzoate ≥97%
Ethyl 3-fluoro-2-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34756448
CAS Number: 2113367-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR