1-Bromo-2-iodo-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

48566-500MG 48566-1G 48566-5G 48566-25G
77327-608EA 181.28 USD
77327-608 77346-850 77363-936 77386-380
1-Bromo-2-iodo-4-isopropylbenzene ≥97%
1-Bromo-2-iodo-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrI
MW: 324.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094020
CAS Number: 1369893-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR