1-Chloro-2-iodo-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

48565-500MG 48565-1G 48565-5G 48565-25G
77327-606EA 277.07 USD
77327-606 77351-220 77363-934 77386-378
1-Chloro-2-iodo-4-isopropylbenzene ≥97%
1-Chloro-2-iodo-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClI
Storage Temperature: Ambient
MDL Number: MFCD29092564
CAS Number: 76662-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR