1,2-diiodo-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

48564-500MG 48564-1G 48564-5G 48564-25G
77327-604EA 236.9 USD
77327-604 77351-218 77363-932 77386-376
1,2-diiodo-4-isopropylbenzene ≥97%
1,2-diiodo-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀I₂
MW: 371.985 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29095359
CAS Number: 1369782-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR