Ethyl 5-chloro-2-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

48557-500MG 48557-1G 48557-5G 48557-25G
77327-596EA 208.06 USD
77327-596 77351-210 77363-924 77386-368
Ethyl 5-chloro-2-fluoro-3-(trifluoromethyl)benzoate ≥97%
Ethyl 5-chloro-2-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28752618
CAS Number: 1805034-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR