5-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48555-500MG 48555-1G 48555-5G
77337-368EA 412 USD
77337-368 77351-208 77375-498
5-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide ≥97%
5-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD33403895
CAS Number: 1512044-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR