5-Bromo-2-fluoro-N-isopropyl-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48554-500MG 48554-1G 48554-5G
77336-034EA 374.92 USD
77336-034 77351-206 77375-496
5-Bromo-2-fluoro-N-isopropyl-3-methylbenzamide ≥97%
5-Bromo-2-fluoro-N-isopropyl-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO
MW: 274.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403894
CAS Number: 1509240-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR