(5-Bromo-2-fluoro-3-methylphenyl)(pyrrolidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48552-500MG 48552-1G 48552-5G
77336-030EA 374.92 USD
77336-030 77351-202 77375-492
(5-Bromo-2-fluoro-3-methylphenyl)(pyrrolidin-1-yl)methanone ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNO
MW: 286.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403892
CAS Number: 2635937-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR