(5-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

48550-500MG 48550-1G 48550-5G
77337-366EA 412 USD
77337-366 77351-198 77375-488
(5-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone ≥97%
(5-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD33403890
CAS Number: 2635937-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR