5-Bromo-2-fluoro-N,N,3-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48547-500MG 48547-1G 48547-5G
77336-028EA 374.92 USD
77336-028 77351-192 77375-482
5-Bromo-2-fluoro-N,N,3-trimethylbenzamide ≥97%
5-Bromo-2-fluoro-N,N,3-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD33403887
CAS Number: 2635937-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR