5-Bromo-2-fluoro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48545-500MG 48545-1G 48545-5G
77336-024EA 374.92 USD
77336-024 77351-188 77375-478
5-Bromo-2-fluoro-3-methylbenzamide ≥97%
5-Bromo-2-fluoro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD28141723
CAS Number: 1503765-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR